Undekan syre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
Hendecanoic acid, Undecylic acid
U0004-25G
U0004-250G
TCIAU0004-25GEA
405
NOK
TCIAU0004-25G
TCIAU0004-250G
Undekan syre ≥98.0% (by GC, titration analysis)
Undekan syre
Formel:
C₁₁H₂₂O₂ MW: 186,29 g/mol Kokepunkt: 228 °C (1013 hPa) Smeltepunkt: 30 °C Tetthet: 0,89 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00002730 CAS-nummer: 112-37-8 EINECS: 203-964-2 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...