1-klorbutan ≥99.0% (by GC)

Leverandør: TCI
Danger

Synonymer: n-Butyl Chloride

C0142-25ML C0142-500ML
TCIAC0142-25MLEA 230 NOK
TCIAC0142-25ML TCIAC0142-500ML
1-klorbutan ≥99.0% (by GC)
1-klorbutan
Formel: CH₃(CH₂)₃Cl
MW: 92,57 g/mol
Kokepunkt: 78°C
Smeltepunkt: –123 °C
Tetthet: 0,8862 g/cm³ (20 °C)
Flash punkt: –13°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001009
CAS-nummer: 109-69-3
EINECS: 203-696-6
FN: 1127
ADR: 3,II
Merck Indeks: 14,01560
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80b837775fb8cf99752f1a054416fd7aa25c2963&vid=98d417ece07d7567bc2348fa8a773aa8fe706cc7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR