1-klortetradekan ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Tetradecyl chloride, Myristyl chloride
C0622-25ML
C0622-500ML
TCIAC0622-25MLEA
220
NOK
TCIAC0622-25ML
TCIAC0622-500ML
1-klortetradekan ≥98.0% (by GC)
1-klortetradekan
Formel:
C₁₄H₂₉Cl MW: 232,84 g/mol Kokepunkt: 155°C Smeltepunkt: 2,2 °C Tetthet: 0,859 g/cm³ (20 °C) Flash punkt: 152°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000958 CAS-nummer: 2425-54-9 EINECS: 219-368-0 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...