(±)-1,2-Pentanediol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
(±)-1,2-Dihydroxy pentane, 1,2-Pentanediol
P1178-25ML
P1178-500ML
TCIAP1178-25MLEA
260
NOK
TCIAP1178-25ML
TCIAP1178-500ML
(±)-1,2-Pentanediol ≥98.0% (by GC)
(±)-1,2-Pentanediol
Formel:
C₅H₁₂O₂ MW: 104,15 g/mol Kokepunkt: 210°C Tetthet: 0,9802 g/cm³ (20 °C) Flash punkt: 104°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00010736 CAS-nummer: 5343-92-0 EINECS: 226-285-3 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...