2,4-Pentanediol ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
2,4-Dihydroxy pentane
P0670-25G
P0670-500G
TCIAP0670-25GEA
660
NOK
TCIAP0670-25G
TCIAP0670-500G
2,4-Pentanediol ≥98.0% (by GC)
2,4-Pentanediol
Formel:
C₅H₁₂O₂ MW: 104,15 g/mol Kokepunkt: 198°C Smeltepunkt: ∼53 °C Tetthet: 0,96 g/cm³ (20 °C) Flash punkt: 101°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004549 CAS-nummer: 625-69-4 EINECS: 210-907-5 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...