1-Oktadekantiol ≥97.0% (by GC, titration analysis)

Leverandør: TCI
Warning

O0005-25G O0005-400G
TCIAO0005-25GEA 325 NOK
TCIAO0005-25G TCIAO0005-400G
1-Oktadekantiol ≥97.0% (by GC, titration analysis)
1-Oktadekantiol
Formel: C₁₈H₃₈S
Kokepunkt: 204 °C (1013 hPa)
Smeltepunkt: 28…31 °C
Tetthet: 0,8475 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004886
CAS-nummer: 2885-00-9
EINECS: 220-744-1
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f04684306df4b8f4e8272da6cb37eaf24ead5e0e&vid=0755e1ffa0e49db104852ccd7088ba407d66f62d&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR