2-Butanethiol ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
1-Methyl-1-propanethiol, 2-Butanethiol, 2-Mercaptobutane, sec-Butyl Mercaptan, sec-Butyl mercaptan
B0686-25ML
B0686-500ML
TCIAB0686-25MLEA
325
NOK
TCIAB0686-25ML
TCIAB0686-500ML
2-Butanethiol ≥97.0% (by GC)
2-Butanethiol
Formel:
C₄H₁₀S MW: 90,19 g/mol Kokepunkt: 84°C Smeltepunkt: –165 °C Tetthet: 0,829 g/cm³ (20 °C) Flash punkt: –23°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004865 CAS-nummer: 513-53-1 EINECS: 208-165-2 FN: 2347 ADR: 3,II Merck Indeks: 14,01578 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...