1H,1H,7H-Dodecafluoro-1-heptanol ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoro-1-heptanol, 1H,1H,7H-Perfluoro-1-heptanol, 1H,1h,7h-dodecafluoro-1-heptanol

D1101-25G D1101-100G
TCIAD1101-25GEA 220 NOK
TCIAD1101-25G TCIAD1101-100G
1H,1H,7H-Dodecafluoro-1-heptanol ≥97.0% (by GC)
1H,1H,7H-Dodecafluoro-1-heptanol
Formel: C₇H₄F₁₂O
MW: 332,09 g/mol
MDL nummer: MFCD00039630
CAS-nummer: 335-99-9
EINECS: 206-406-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=41852aa1d68798cdc62a12817bbc1d3aec8b1acd&vid=41852aa1d68798cdc62a12817bbc1d3aec8b1acd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR