trans-2-Heksenal ≥97.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
(E)-2-Hexenal, trans-2-Hexenal
H0345-25ML
H0345-500ML
H0345-100ML
TCIAH0345-25MLEA
230
NOK
TCIAH0345-25ML
TCIAH0345-500ML
TCIAH0345-100ML
trans-2-Heksenal ≥97.0% (by GC)
trans-2-Heksenal
Formel:
C₆H₁₀O MW: 98,14 g/mol Kokepunkt: 146°C Tetthet: 0,846 g/cm³ (20 °C) Flash punkt: 43°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00007008 CAS-nummer: 6728-26-3 EINECS: 229-778-1 FN: 1988 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...