3-Kloropropen ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
3-Chloro-1-propene, Allyl chloride
C0274-25ML
C0274-500ML
TCIAC0274-25MLEA
220
NOK
TCIAC0274-25ML
TCIAC0274-500ML
3-Kloropropen ≥98.0% (by GC)
3-Kloropropen
Formel:
C₃H₅Cl MW: 76,53 g/mol Kokepunkt: 45°C Smeltepunkt: –136 °C Tetthet: 0,93 g/cm³ (20 °C) Flash punkt: –29°C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00000984 CAS-nummer: 107-05-1 EINECS: 203-457-6 FN: 1100 ADR: 3,I Merck Indeks: 14,00289 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...