trans-2-Metyl-2-butenal ≥95.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Trans-2-methyl-2-butenal, Tiglic aldehyde, (E)-2-Methyl-2-butenal

T1003-25ML T1003-5ML
TCIAT1003-25MLEA 1325 NOK
TCIAT1003-25ML TCIAT1003-5ML
trans-2-Metyl-2-butenal ≥95.0% (by GC)
trans-2-Metyl-2-butenal
Formel: C₅H₈O
MW: 84,12 g/mol
Kokepunkt: 116°C
Tetthet: 0,8688 g/cm³ (20 °C)
Flash punkt: 18°C
MDL nummer: MFCD00006977
CAS-nummer: 497-03-0
EINECS: 207-833-0
FN: 1989
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba99409d6c96c5761fb9ba051dc19ea2f6e16b49&vid=8eff4c299086ec975e0c1a6d9bb02e5dfbf75e11&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR