Methyl-p-anisate ≥99.0% (by GC)

Leverandør: TCI

Synonymer: 4-Methoxybenzoic acid methyl ester, 4-Anisic acid methyl ester, p-Anisic acid methyl ester, Methyl 4-methoxybenzoate

A1125-25G A1125-500G A1125-100G
TCIAA1125-25GEA 230 NOK
TCIAA1125-25G TCIAA1125-500G TCIAA1125-100G
Methyl-p-anisate ≥99.0% (by GC)
Methyl-p-anisate
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokepunkt: 255°C
MDL nummer: MFCD00008437
CAS-nummer: 121-98-2
EINECS: 204-513-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e297c287b1cdc37853140693050c9b603a81ecb7&vid=e297c287b1cdc37853140693050c9b603a81ecb7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 99.0 %
Melting point 47.0 to 51.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR