α-Cyclopropylbenzyl alcohol ≥96.0% (by GC)

Leverandør: TCI

Synonymer: alpha-Cyclopropylbenzyl alcohol, Cyclopropyl(phenyl)methanol, α-Cyclopropylbenzyl alcohol, Cyclopropyl phenyl carbinol, Cyclopropylphenylcarbinol

C0986-25ML C0986-5ML
TCIAC0986-25MLEA 3750 NOK
TCIAC0986-25ML TCIAC0986-5ML
α-Cyclopropylbenzyl alcohol ≥96.0% (by GC)
α-Cyclopropylbenzyl alcohol
Formel: C₁₀H₁₂O
MW: 148,2 g/mol
Kokepunkt: 132°C
Flash punkt: 112°C
MDL nummer: MFCD00001299
CAS-nummer: 1007-03-0
EINECS: 213-749-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=def6bfdb07e0c93c77e17e941d54667ac15d80e3&vid=def6bfdb07e0c93c77e17e941d54667ac15d80e3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR