5-Bromo-o-anisaldehyde ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Bromo-2-methoxybenzaldehyde, 5-Bromo-2-anisaldehyde

B1791-25G B1791-250G
TCIAB1791-25GEA 555 NOK
TCIAB1791-25G TCIAB1791-250G
5-Bromo-o-anisaldehyde ≥98.0% (by GC)
5-Bromo-o-anisaldehyde
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
Kokepunkt: 269 °C (1013 hPa)
Smeltepunkt: 117 °C
MDL nummer: MFCD00003347
CAS-nummer: 25016-01-7
EINECS: 246-564-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a8a4791a64a9b6113a55e21fe61482df7630f88&vid=79fd789cca91eb84752b97a051bfc9e53ff9b9c3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR