4-Bromo-3-chloroaniline ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
B3709-25G B3709-5G
TCIAB3709-25GEA 3850 NOK
TCIAB3709-25G TCIAB3709-5G
4-Bromo-3-chloroaniline ≥98.0% (by GC, titration analysis)
4-Bromo-3-chloroaniline
Formel: C₆H₅BrClN
MW: 206,47 g/mol
Kokepunkt: 264 °C (1013 hPa)
Smeltepunkt: 67…68 °C
MDL nummer: MFCD00041311
CAS-nummer: 21402-26-6
EINECS: 244-370-3
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32bdd5a9a25436c743fb2db578e5e90bcb7b2e4c&vid=8346432f9e1d7f0fb76cb06b3ff1f5c9b1ee5498&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C