4-Bromo-2-chlorophenol ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
B1819-25G
B1819-5G
TCIAB1819-25GEA
650
NOK
TCIAB1819-25G
TCIAB1819-5G
4-Bromo-2-chlorophenol ≥97.0% (by GC)
4-Bromo-2-chlorophenol
Formel:
C₆H₄BrClO MW: 207,45 g/mol Kokepunkt: 234°C Smeltepunkt: 50…51 °C Tetthet: 1,617 g/cm³ (20 °C) |
MDL nummer:
MFCD00002166 CAS-nummer: 3964-56-5 EINECS: 223-572-5 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 48.0 to 51.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...