1-Bromo-2,3-difluorobenzene ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2,3-Difluorobromobenzene
B1800-1G
B1800-5G
B1800-25G
TCIAB1800-1GEA
300
NOK
TCIAB1800-1G
TCIAB1800-5G
TCIAB1800-25G
1-Bromo-2,3-difluorobenzene ≥98.0% (by GC)
1-Bromo-2,3-difluorobenzene
Formel:
C₆H₃BrF₂ MW: 192,99 g/mol Kokepunkt: 234 °C (765 -rr) Tetthet: 1,72 g/cm³ (20 °C) Flash punkt: 61°C |
MDL nummer:
MFCD00061136 CAS-nummer: 38573-88-5 |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...