1-Bromo-2,4,5-trifluorobenzene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,4,5-Trifluorobromobenzene

B1167-25G B1167-5G
TCIAB1167-25GEA 1125 NOK
TCIAB1167-25G TCIAB1167-5G
1-Bromo-2,4,5-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,4,5-trifluorobenzene
Formel: C₆H₂BrF₃
MW: 210,98 g/mol
Kokepunkt: 144°C
Smeltepunkt: –19 °C
Tetthet: 1,8022 g/cm³ (20 °C)
Flash punkt: 55°C
MDL nummer: MFCD00000306
CAS-nummer: 327-52-6
EINECS: 206-318-8
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7345460e6bda852af9b14bc3ca56d50be2e70a&vid=f04bacfc892dccbafb03325e4d933df54877c165&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR