1-Bromo-2,4,6-trifluorobenzene ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2-Bromo-1,3,5-trifluorobenzene

B1168-25G B1168-5G
TCIAB1168-25GEA 2875 NOK
TCIAB1168-25G TCIAB1168-5G
1-Bromo-2,4,6-trifluorobenzene ≥98.0% (by GC)
1-Bromo-2,4,6-trifluorobenzene
Formel: C₆H₂BrF₃
MW: 210,98 g/mol
Kokepunkt: 140°C
Smeltepunkt: 3,5 °C
Tetthet: 1,79 g/cm³ (20 °C)
Flash punkt: 58°C
MDL nummer: MFCD00000334
CAS-nummer: 2367-76-2
EINECS: 219-125-9
FN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5680e6379dd1d5346ccebc805a64999050ed2a58&vid=1187cb8fadee49ba8c8103cd6da4e6be68a9ea06&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR