Bis(4-hydroxyphenyl)sulphone ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 4,4'-Sulphonyldiphenol, 4,4'-Dihydroxy diphenyl sulphone, 4-Hydroxyphenyl sulphone, 4,4'-sulphonyldiphenol
B0495-25G B0495-500G
TCIAB0495-25GEA 220 NOK
TCIAB0495-25G TCIAB0495-500G
Bis(4-hydroxyphenyl)sulphone ≥98.0% (by HPLC, titration analysis)
Bis(4-hydroxyphenyl)sulphone
Formel: C₁₂H₁₀O₄S
MW: 250,28 g/mol
Smeltepunkt: 247°C
MDL nummer: MFCD00002350
CAS-nummer: 80-09-1
EINECS: 201-250-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f730bdfad5cc46d964d0b1248e999844ef9fdf62&vid=f730bdfad5cc46d964d0b1248e999844ef9fdf62&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency