Tetramethylene sulphoxide ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
Tetrahydrothiophene 1-oxide
T0146-25G
T0146-500G
TCIAT0146-25GEA
825
NOK
TCIAT0146-25G
TCIAT0146-500G
Tetramethylene sulphoxide ≥98.0% (by GC)
Tetramethylene sulphoxide
Formel:
C₄H₈OS MW: 104,17 g/mol Kokepunkt: 107°C |
MDL nummer:
MFCD00005477 CAS-nummer: 1600-44-8 EINECS: 216-493-2 |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...