3,5-Dichlorophenylhydrazinium chloride ≥90.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride
D2644-25G D2644-5G
TCIAD2644-25GEA 1575 NOK
TCIAD2644-25G TCIAD2644-5G
3,5-Dichlorophenylhydrazinium chloride ≥90.0% (by HPLC, titration analysis)
3,5-Dichlorophenylhydrazinium chloride
Formel: C₆H₇Cl₃N₂
MW: 213,49 g/mol
MDL nummer: MFCD00012938
CAS-nummer: 63352-99-8
EINECS: 264-109-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c2c5523856c89751c886097b1a51afe3aaaa5a5&vid=2c2c5523856c89751c886097b1a51afe3aaaa5a5&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Purity (HPLC) min. 90.0 area%
Purity (Potassium iodate Method) min. 90.0 %