Benzophenone imine ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Benzophenoneimine
B1912-25G
B1912-5G
TCIAB1912-25GEA
1050
NOK
TCIAB1912-25G
TCIAB1912-5G
Benzophenone imine ≥95.0% (by GC)
Benzophenone imine
Formel:
C₁₃H₁₁N MW: 181,24 g/mol Kokepunkt: 108°C |
MDL nummer:
MFCD00001760 CAS-nummer: 1013-88-3 |
Spesifikasjon Testresultater
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...