4-(4-Nitrobenzyl)pyridine ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

Synonymer: 4-(p-Nitrobenzyl)Pyridine

N0183-1G N0183-25G
TCIAN0183-1GEA 220 NOK
TCIAN0183-1G TCIAN0183-25G
4-(4-Nitrobenzyl)pyridine ≥98.0% (by titrimetric analysis)
4-(4-Nitrobenzyl)pyridine
Formel: C₁₂H₁₀N₂O₂
MW: 214,22 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006445
CAS-nummer: 1083-48-3
EINECS: 214-108-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15479a3b42d363670d300ad8c9c6b922b4391ad3&vid=15479a3b42d363670d300ad8c9c6b922b4391ad3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 72.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR