5-Brom-2-fluorbensaldehyd ≥95.0% (by GC)
Leverandør: TCI
|
Warning
|
B1789-25G
B1789-5G
TCIAB1789-25GEA
865
NOK
TCIAB1789-25G
TCIAB1789-5G
5-Brom-2-fluorbensaldehyd ≥95.0% (by GC)
5-Brom-2-fluorbensaldehyd
Formel:
C₇H₄BrFO MW: 203,01 g/mol Kokepunkt: 229 °C (1013 hPa) Smeltepunkt: 49 °C Tetthet: 1,71 g/cm³ (20 °C) Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00070755 CAS-nummer: 93777-26-5 EINECS: 298-056-6 |
Spesifikasjon Testresultater
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...