4-Bromanilin ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
B1393-25G
B1393-500G
B1393-100G
TCIAB1393-25GEA
260
NOK
TCIAB1393-25G
TCIAB1393-500G
TCIAB1393-100G
4-Bromanilin ≥99.0% (by GC)
4-Bromanilin
Formel:
C₆H₆BrN MW: 172,02 g/mol Kokepunkt: 124°C Smeltepunkt: 62…64 °C Tetthet: 1,799 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007822 CAS-nummer: 106-40-1 EINECS: 203-393-9 ADR: 6.1,III Merck Indeks: 14,01404 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Solubility in Toluene | almost transparency |
Melting point | 63.0 to 67.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...