4-Nitrobensaldehyd ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
N0559-25G
N0559-500G
N0559-100G
TCIAN0559-25GEA
350
NOK
TCIAN0559-25G
TCIAN0559-500G
TCIAN0559-100G
4-Nitrobensaldehyd ≥98.0% (by GC)
4-Nitrobensaldehyd
Formel:
C₇H₅NO₃ Kokepunkt: 300 °C (1013 hPa) Smeltepunkt: 106 °C Tetthet: 1,546 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007346 CAS-nummer: 555-16-8 EINECS: 209-084-5 Merck Indeks: 14,06587 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 102.0 to 107.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...