5-Nitrovanillin ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
4-Hydroxy-3-methoxy-5-nitrobenzaldehyde
H0728-25G
H0728-5G
TCIAH0728-25GEA
865
NOK
TCIAH0728-25G
TCIAH0728-5G
5-Nitrovanillin ≥98.0% (by titrimetric analysis)
5-Nitrovanillin
Formel:
C₈H₇NO₅ MW: 197,15 g/mol Smeltepunkt: 175…178 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007118 CAS-nummer: 6635-20-7 EINECS: 229-633-2 |
Spesifikasjon Testresultater
Appearance | Yellow to Brown to Dark green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 174.0 to 178.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...