3-Nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Hydroxy-3-nitrobenzaldehyde
N0823-1G
N0823-5G
TCIAN0823-1GEA
1050
NOK
TCIAN0823-1G
TCIAN0823-5G
3-Nitrosalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Nitrosalicylaldehyde
Formel:
C₇H₅NO₄ MW: 167,12 g/mol |
MDL nummer:
MFCD00041874 CAS-nummer: 5274-70-4 EINECS: 226-098-7 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...