3,5-Dimetoksybenzaldehyd ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
D1164-25G
D1164-5G
TCIAD1164-25GEA
1525
NOK
TCIAD1164-25G
TCIAD1164-5G
3,5-Dimetoksybenzaldehyd ≥98.0% (by GC)
3,5-Dimetoksybenzaldehyd
Formel:
C₉H₁₀O₃ MW: 166,18 g/mol Kokepunkt: 151°C Smeltepunkt: 48 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00003366 CAS-nummer: 7311-34-4 EINECS: 230-772-6 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 48.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...