Benzonitril ≥99.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Cyanobenzene, Phenylcyanide

B0082-25G B0082-500G
TCIAB0082-25GEA 245 NOK
TCIAB0082-25G TCIAB0082-500G
Benzonitril ≥99.0% (by GC)
Benzonitril
Formel: C₇H₅N
MW: 103,12 g/mol
Kokepunkt: 191°C
Smeltepunkt: –13 °C
Tetthet: 1 g/cm³ (20 °C)
Flash punkt: 75°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001770
CAS-nummer: 100-47-0
EINECS: 202-855-7
FN: 2224
ADR: 6.1,II
Merck Indeks: 14,01097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=307aa380e236620f66f76c3b786dc99243cb8eac&vid=6509aa01f5b2e75b196a7a2b5b96439d80c9a1bd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR