3-Hydroksybensonitril ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

C1583-25G C1583-5G
TCIAC1583-25GEA 1350 NOK
TCIAC1583-25G TCIAC1583-5G
3-Hydroksybensonitril ≥98.0% (by GC)
3-Hydroksybensonitril
Formel: C₇H₅NO
MW: 119,12 g/mol
Kokepunkt: 265 °C (1013 hPa)
Smeltepunkt: 84,5…86 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002252
CAS-nummer: 873-62-1
EINECS: 212-845-4
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20d6c4cd264da1a0718db13e2c1f63c6372fa80c&vid=49d1bfa8bd92fab172665764989594539be25c30&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystaline
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR