Heksyl benzoat ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Benzoic acid hexyl ester

B1076-25ML B1076-500ML
TCIAB1076-25MLEA 220 NOK
TCIAB1076-25ML TCIAB1076-500ML
Heksyl benzoat ≥98.0% (by GC)
Heksyl benzoat
Formel: C₁₃H₁₈O₂
MW: 206,28 g/mol
Kokepunkt: 272°C
Tetthet: 0,9992 g/cm³ (0 °C)
Flash punkt: 148°C
MDL nummer: MFCD00051714
CAS-nummer: 6789-88-4
EINECS: 229-856-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6728877c5b01c60d4928e91527c1e5133666c4c&vid=6b115a2df4a749be55c532747630d7391b214171&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR