2-Etoksybensoe syre ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

E0044-25G E0044-500G
TCIAE0044-25GEA 340 NOK
TCIAE0044-25G TCIAE0044-500G
2-Etoksybensoe syre ≥98.0% (by GC, titration analysis)
2-Etoksybensoe syre
Formel: C₉H₁₀O₃
MW: 166,18 g/mol
Kokepunkt: 301 °C (1013 hPa)
Smeltepunkt: 25 °C
Tetthet: 1,173 g/cm³ (20 °C)
MDL nummer: MFCD00002438
CAS-nummer: 134-11-2
EINECS: 205-130-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c9f338ee73cb1dde4ffcbfe278ae028d555b924&vid=47ca9c3ab91c6ae3862d7274ba2aa2189ab7c462&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR