2,3,4-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
T1116-25G
T1116-5G
TCIAT1116-25GEA
1850
NOK
TCIAT1116-25G
TCIAT1116-5G
2,3,4-Trimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,3,4-Trimethoxybenzoic acid
Formel:
C₁₀H₁₂O₅ MW: 212,2 g/mol Kokepunkt: 110 - 115 °C (0,04 -rr) Smeltepunkt: 104 °C |
MDL nummer:
MFCD00002433 CAS-nummer: 573-11-5 EINECS: 209-350-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 100.0 to 103.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...