Trimellitic acid ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid
B0042-500G B0042-25G B0042-100G
TCIAB0042-500GEA 3125 NOK
TCIAB0042-500G TCIAB0042-25G TCIAB0042-100G
Trimellitic acid ≥98.0% (by HPLC, titration analysis)
Trimellitic acid
Formel: C₉H₆O₆
MW: 210,14 g/mol
Kokepunkt: 107 - 113 °C (0,1 -rr)
Smeltepunkt: 240…245 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002470
CAS-nummer: 528-44-9
EINECS: 208-432-3
Merck Indeks: 14,09702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce66f8880bb7a4cf274f44a5a9bb0cf8575aaa95&vid=635d9d20b902f942abcfdc002f026c4110d64269&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency