Trimesic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: Benzene-1,3,5-tricarboxylic acid, 1,3,5-Benzenetricarboxylic acid
B0043-25G B0043-500G B0043-100G
TCIAB0043-25GEA 270 NOK
TCIAB0043-25G TCIAB0043-500G TCIAB0043-100G
Trimesic acid ≥98.0% (by titrimetric analysis)
Trimesic acid
Formel: C₉H₆O₆
MW: 210,14 g/mol
Smeltepunkt: 345…350 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002517
CAS-nummer: 554-95-0
EINECS: 209-077-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8daa1c3e161c074e28132e98f2035fc9c695e4dd&vid=ab728e4c7eeb64390636b8be18949ecfec30aefe&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol very faint turbidity