1,2,4,5-Tetraklorbensen ≥99.0% (by GC)
Leverandør: TCI
|
Danger
|
T0060-25G
T0060-500G
TCIAT0060-25GEA
230
NOK
TCIAT0060-25G
TCIAT0060-500G
1,2,4,5-Tetraklorbensen ≥99.0% (by GC)
1,2,4,5-Tetraklorbensen
Formel:
C₆H₂Cl₄ MW: 215,89 g/mol Kokepunkt: 246°C Smeltepunkt: 136 °C Tetthet: 1,732 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00000549 CAS-nummer: 95-94-3 EINECS: 202-466-2 FN: 3077 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 140.0 to 143.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...