3-Klorbensaldehyd ≥98.0% (by GC)

Leverandør: TCI
Warning

C0124-25G C0124-500G C0124-100G
TCIAC0124-25GEA 220 NOK
TCIAC0124-25G TCIAC0124-500G TCIAC0124-100G
3-Klorbensaldehyd ≥98.0% (by GC)
3-Klorbensaldehyd
Formel: C₇H₅ClO
Kokepunkt: 214°C
Smeltepunkt: 17…18 °C
Tetthet: 1,241 g/cm³ (20 °C)
Flash punkt: 88°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003350
CAS-nummer: 587-04-2
EINECS: 209-596-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05f5be99a15f4f563e72257b0f838cc1f3a68a05&vid=125fbb7bf2db3e2e9af6ae0cd24919d4e2f1044b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR