3,5-Diklorbensonitril ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
D3261-25G
D3261-5G
TCIAD3261-25GEA
1500
NOK
TCIAD3261-25G
TCIAD3261-5G
3,5-Diklorbensonitril ≥98.0% (by GC)
3,5-Diklorbensonitril
Formel:
C₇H₃Cl₂N MW: 172,01 g/mol Kokepunkt: 269 °C (1013 hPa) Smeltepunkt: 66 °C |
MDL nummer:
MFCD00001800 CAS-nummer: 6575-00-4 EINECS: 229-495-3 FN: 3439 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 69.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...