4-klor-3-nitrobensonitril ≥98.0% (by GC)

Leverandør: TCI
Warning

C1548-25G C1548-5G
TCIAC1548-25GEA 1075 NOK
TCIAC1548-25G TCIAC1548-5G
4-klor-3-nitrobensonitril ≥98.0% (by GC)
4-klor-3-nitrobensonitril
Formel: C₇H₃ClN₂O₂
MW: 182,57 g/mol
Smeltepunkt: 100 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016987
CAS-nummer: 939-80-0
EINECS: 213-364-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a58331d7aab78cbd86f674f89732482ac3609d5&vid=5af5e8438d7d2e8bcaa986c6dfc37cf23e4fbdcb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR