2,5-Diklorbenso syre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
D0338-100G D0338-25G
TCIAD0338-100GEA 985 NOK
TCIAD0338-100G TCIAD0338-25G
2,5-Diklorbenso syre ≥98.0% (by GC, titration analysis)
2,5-Diklorbenso syre
Formel: C₇H₄Cl₂O₂
MW: 191,01 g/mol
Kokepunkt: 301 °C (1013 hPa)
Smeltepunkt: 151…154 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002416
CAS-nummer: 50-79-3
EINECS: 200-065-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=562db9628a40c492eb3d1058d6abbe255f8d569a&vid=86ca7b767814ed61b706de9bff791bfc59fac23e&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 152.0 to 156.0 deg-C