2,6-Diklorbenso syre ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
D0339-25G D0339-5G
TCIAD0339-25GEA 350 NOK
TCIAD0339-25G TCIAD0339-5G
2,6-Diklorbenso syre ≥98.0% (by GC, titration analysis)
2,6-Diklorbenso syre
Formel: C₇H₄Cl₂O₂
MW: 191,01 g/mol
Kokepunkt: 299 °C (1013 hPa)
Smeltepunkt: 138…139 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002418
CAS-nummer: 50-30-6
EINECS: 200-025-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eb271c16c667af883675452ff0425e773888e1f&vid=18d01ae0fdb122e6ebc08a8c5d3e4b1fcd21d5a7&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 145.0 deg-C
Solubility in Methanol almost transparency