4-Amino-2-chlorobenzoic acid ≥97.0% (by titrimetric analysis)
Leverandør: TCI
Warning
A0951-25G A0951-5G
TCIAA0951-25GEA 1350 NOK
TCIAA0951-25G TCIAA0951-5G
4-Amino-2-chlorobenzoic acid ≥97.0% (by titrimetric analysis)
4-Amino-2-chlorobenzoic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Kokepunkt: 330 °C (1013 hPa)
Smeltepunkt: 220 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007772
CAS-nummer: 2457-76-3
EINECS: 219-540-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c68d60bbd24758daa710fc1c28e531b1329e829a&vid=f4c9e37ce997a32721daf4e263fc4db8c88cb17f&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Light red to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Melting point 206.0 to 216.0 deg-C
Solubility in hot Methanol almost transparency