4-klor-2-fluorbensaldehyd ≥98.0% (by GC)

Leverandør: TCI
Warning

C1597-25G C1597-5G
TCIAC1597-25GEA 1950 NOK
TCIAC1597-25G TCIAC1597-5G
4-klor-2-fluorbensaldehyd ≥98.0% (by GC)
4-klor-2-fluorbensaldehyd
Formel: C₇H₄ClFO
MW: 158,56 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00143282
CAS-nummer: 61072-56-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba071de1f3c9fca27d2cb539aff8b9a215f452ae&vid=a935f654c9dbca28350cee0791bbcc3b232eabd7&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 61.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR