2,4-Dichloro-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
D2386-25G D2386-5G
TCIAD2386-25GEA 1825 NOK
TCIAD2386-25G TCIAD2386-5G
2,4-Dichloro-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,4-Dichloro-5-fluorobenzoic acid
Formel: C₇H₃Cl₂FO₂
MW: 209 g/mol
Kokepunkt: 298 °C (1013 hPa)
Smeltepunkt: 141…144 °C
MDL nummer: MFCD00075281
CAS-nummer: 86522-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afea9334bd72b0307e46c128b0e0ce886e21c13e&vid=5b4a0e98c6bbc9d6b639d7ad2e59413de9ff4dc9&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C