3-Chloro-2,4,5-trifluorobenzoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2,4,5-Trifluoro-3-chlorobenzoic acid

C2261-25G C2261-5G
TCIAC2261-25GEA 1575 NOK
TCIAC2261-25G TCIAC2261-5G
3-Chloro-2,4,5-trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
3-Chloro-2,4,5-trifluorobenzoic acid
Formel: C₇H₂ClF₃O₂
MW: 210,54 g/mol
MDL nummer: MFCD00153101
CAS-nummer: 101513-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16ce6e9021ee21d1fe308fa53fe628869279e380&vid=4525e48800891aac69ce85bf118f3a018255902a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 112.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR