4-Klor-2-fluor-1-nitrobensen ≥96.0% (by GC)

Leverandør: TCI

Synonymer: 4-Chloro-2-fluoronitrobenzene

C1261-25G C1261-5G
TCIAC1261-25GEA 825 NOK
TCIAC1261-25G TCIAC1261-5G
4-Klor-2-fluor-1-nitrobensen ≥96.0% (by GC)
4-Klor-2-fluor-1-nitrobensen
Formel: C₆H₃ClFNO₂
MW: 175,55 g/mol
MDL nummer: MFCD00042211
CAS-nummer: 700-37-8
EINECS: 211-842-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a4e94c3abe1f93030c59667f0e0e474ae3c73f6&vid=afe550359e00971cad0d418bea419e03cadfc7df&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 47.0 to 50.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR