2,3,4,5,6-Pentachloroaniline ≥97.0% (by GC)

Leverandør: TCI
Danger

Synonymer: Pentachloroaniline

P0917-1G P0917-5G
TCIAP0917-1GEA 1025 NOK
TCIAP0917-1G TCIAP0917-5G
2,3,4,5,6-Pentachloroaniline ≥97.0% (by GC)
2,3,4,5,6-Pentachloroaniline
Formel: C₆H₂Cl₅N
MW: 265,35 g/mol
Kokepunkt: 321 °C (1013 hPa)
Smeltepunkt: 206 °C
MDL nummer: MFCD00014769
CAS-nummer: 527-20-8
EINECS: 208-410-3
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6db78951693bae2b6fb44ed715c785471280d258&vid=bc0bfaeff6c519d50d05f600d66c1ba9a9fd1259&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 232.0 to 237.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR