1-klor-4-nitrobenzen ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-Chloronitrobenzene

C0222-25G C0222-500G
TCIAC0222-25GEA 245 NOK
TCIAC0222-25G TCIAC0222-500G
1-klor-4-nitrobenzen ≥98.0% (by GC)
1-klor-4-nitrobenzen
Formel: C₆H₄ClNO₂
MW: 157,56 g/mol
Kokepunkt: 242°C
Smeltepunkt: 83 °C
Tetthet: 1,52 g/cm³ (20 °C)
Flash punkt: 127 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007285
CAS-nummer: 100-00-5
EINECS: 202-809-6
FN: 1578
ADR: 6.1,II
Merck Indeks: 14,02151
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d0238a95542d82cc17d690625fcac66c804e611&vid=977036f60436cea4d6b5024ff7297c84fb70228a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 82.0 to 86.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR